siRNA Details
| Gene Symbol: ALDH1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034173.3 | NP_001029345.2 | 15 | 1970 | ||
| NR_027752.1 | 15 | 1966 | ||||
| XM_011537986.2 | XP_011536288.1 | 13 | 1529 | |||
| XM_011537988.2 | XP_011536290.1 | 15 | 1991 | |||
| XM_017018889.1 | XP_016874378.1 | 16 | 2176 | |||
| GenBank mRNA | AK095827.1 | - | 15 | 1865 | ||
| AK300373.1 | - | 15 | 1865 | |||
| BC103934.1 | - | 15 | 1865 | 40008174 | ||
| BC103935.1 | - | 15 | 1861 | 40008176 | ||
| CR627287.1 | - | 11 | 1834 | |||

ALDH1L2
Genomic Map