siRNA Details
| Gene Symbol: GRASP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271856.1 | NP_001258785.1 | 6 | 496 | ||
| NM_181711.3 | NP_859062.1 | 7 | 730 | |||
| XM_005268691.3 | XP_005268748.1 | 7 | 1898 | |||
| XM_011537996.1 | XP_011536298.1 | 6 | 1676 | |||
| GenBank mRNA | BC035500.2 | - | 6 | 467 | 4422992 | |
| BC052967.1 | - | 6 | 414 | |||
| BC053666.1 | - | 7 | 711 | 6151168 | ||

GRASP
Genomic Map