siRNA Details
| Gene Symbol: FAM71D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173526.3 | NP_775797.2 | 6 | 1296 | ||
| XM_011536500.1 | XP_011534802.1 | 6 | 1260 | |||
| XM_011536502.1 | XP_011534804.1 | 6 | 1195 | |||
| XM_011536503.2 | XP_011534805.1 | 5 | 2237 | |||
| GenBank mRNA | AK093449.1 | - | 6 | 1260 | ||

FAM71D
Genomic Map