siRNA Details
| Gene Symbol: SYNE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152592.3 | NP_689805.3 | 6 | 1225 | ||
| XM_005267377.3 | XP_005267434.1 | 7 | 1322 | |||
| XM_006720063.3 | XP_006720126.1 | 7 | 1422 | |||
| XM_011536515.2 | XP_011534817.1 | 7 | 1357 | |||
| XM_017021056.1 | XP_016876545.1 | 6 | 1224 | |||
| XM_017021057.1 | XP_016876546.1 | 7 | 1322 | |||
| GenBank mRNA | AK131436.1 | - | 7 | 1090 | ||
| BC146604.1 | - | 7 | 1225 | 40123293 | ||
| BX248076.1 | - | 6 | 1225 | |||
| BX342493.1 | - | 3 | 527 | |||
| BX648302.1 | - | 5 | 956 | |||

SYNE3
Genomic Map