siRNA Details
| Gene Symbol: TMEM30B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017970.2 | NP_001017970.1 | 1 | 2069 | ||
| GenBank mRNA | AK091169.1 | - | 1 | 1498 | ||
| DA431345.1 | - | 1 | 76 | |||
| DA666826.1 | - | 1 | 388 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM30B| Gene Symbol: TMEM30B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017970.2 | NP_001017970.1 | 1 | 2069 | ||
| GenBank mRNA | AK091169.1 | - | 1 | 1498 | ||
| DA431345.1 | - | 1 | 76 | |||
| DA666826.1 | - | 1 | 388 | |||