siRNA Details
| Gene Symbol: TMEM30B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017970.2 | NP_001017970.1 | 1 | 714 | ||
| GenBank mRNA | AK091169.1 | - | 1 | 143 | ||
| BC101726.1 | - | 1 | 95 | 8069232 | ||
| BC113559.1 | - | 1 | 95 | 8322611 | ||
| DB237714.1 | - | 1 | 240 | |||

TMEM30B
Genomic Map