siRNA Details
| Gene Symbol: MDGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113498.2 | NP_001106970.3 | 13 | 2726 | ||
| NM_182830.4 | NP_878250.2 | 13 | 2432 | |||
| XM_011536519.2 | XP_011534821.1 | 13 | 2581 | |||
| XM_011536525.2 | XP_011534827.1 | 10 | 1863 | |||
| XM_017021060.1 | XP_016876549.1 | 9 | 1692 | |||
| GenBank mRNA | AK123307.1 | - | 2 | 276 | ||
| AK309211.1 | - | 13 | 2729 | |||
| AY328482.1 | - | 7 | 1359 | |||
| AY358125.1 | - | 12 | 2306 | |||
| AY369208.1 | - | 13 | 2522 | |||

MDGA2
Genomic Map