siRNA Details
| Gene Symbol: ZFPM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153813.2 | NP_722520.2 | 2, 3 | 179 | ||
| XM_011522912.2 | XP_011521214.1 | 2, 3 | 4028 | |||
| XM_011522914.2 | XP_011521216.1 | 2, 3 | 263 | |||
| GenBank mRNA | AF488691.1 | - | 2, 3 | 468 | ||
| BE219988.1 | - | 2, 3 | 179 | |||

ZFPM1
Genomic Map