siRNA Details
| Gene Symbol: ANKFN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153228.2 | NP_694960.2 | 10 | 1156 | ||
| XM_006721728.3 | XP_006721791.1 | 9 | 1379 | |||
| XM_011524428.2 | XP_011522730.1 | 10 | 1156 | |||
| XM_011524429.2 | XP_011522731.1 | 10 | 1313 | |||
| XM_011524430.2 | XP_011522732.1 | 11 | 1279 | |||
| XM_011524431.2 | XP_011522733.1 | 10 | 1211 | |||
| XM_011524433.2 | XP_011522735.1 | 4 | 569 | |||
| XM_017024263.1 | XP_016879752.1 | 12 | 1373 | |||
| XM_017024264.1 | XP_016879753.1 | 12 | 2358 | |||
| XM_017024265.1 | XP_016879754.1 | 12 | 1567 | |||
| XM_017024266.1 | XP_016879755.1 | 10 | 1328 | |||
| XM_017024267.1 | XP_016879756.1 | 9 | 1180 | |||
| XM_017024268.1 | XP_016879757.1 | 11 | 2264 | |||
| XM_017024269.1 | XP_016879758.1 | 9 | 1453 | |||
| XM_017024270.1 | XP_016879759.1 | 12 | 1371 | |||
| GenBank mRNA | AK095654.1 | - | 10 | 1156 | ||

ANKFN1
Genomic Map