siRNA Details
| Gene Symbol: TMEM92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168215.1 | NP_001161687.1 | 6 | 715 | ||
| NM_153229.2 | NP_694961.2 | 5 | 680 | |||
| XM_006721730.3 | XP_006721793.1 | 5 | 638 | |||
| GenBank mRNA | AK090637.1 | - | 5 | 635 | ||
| AY358204.1 | - | 6 | 715 | |||
| BC064507.1 | - | 5 | 608 | 6455254 | ||

TMEM92
Genomic Map