siRNA Details
| Gene Symbol: RHBDL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138328.2 | NP_612201.1 | 2 | 138 | ||
| XM_017024272.1 | XP_016879761.1 | 2 | 143 | |||
| XM_017024275.1 | XP_016879764.1 | 2 | 143 | |||
| XM_017024279.1 | XP_016879768.1 | 1 | 940 | |||
| GenBank mRNA | AJ313480.1 | - | 2 | 138 | ||
| AK124570.1 | - | 1 | 111 | |||
| BC100975.1 | - | 2 | 137 | 40005244 | ||
| BC100976.1 | - | 2 | 138 | |||
| BC100978.1 | - | 2 | 138 | 40005247 | ||

RHBDL3
Genomic Map