siRNA Details
| Gene Symbol: ZNF600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321866.1 | NP_001308795.1 | 6 | 1899 | ||
| NM_001321867.1 | NP_001308796.1 | 4 | 1276 | |||
| NM_198457.3 | NP_940859.3 | 3 | 1149 | |||
| XM_017026390.1 | XP_016881879.1 | 7 | 1826 | |||
| XM_017026391.1 | XP_016881880.1 | 6 | 1802 | |||
| XM_017026392.1 | XP_016881881.1 | 6 | 1600 | |||
| GenBank mRNA | AK075230.1 | - | 1 | 436 | ||
| AK131231.1 | - | 3 | 1139 | |||
| BX640933.1 | - | 3 | 1121 | |||

ZNF600
Genomic Map