siRNA Details
| Gene Symbol: ZNF600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321866.1 | NP_001308795.1 | 6 | 1353 | ||
| NM_001321867.1 | NP_001308796.1 | 4 | 730 | |||
| NM_198457.3 | NP_940859.3 | 3 | 603 | |||
| XM_017026390.1 | XP_016881879.1 | 7 | 1280 | |||
| XM_017026391.1 | XP_016881880.1 | 6 | 1256 | |||
| XM_017026392.1 | XP_016881881.1 | 6 | 1054 | |||
| GenBank mRNA | AK131231.1 | - | 3 | 593 | ||
| BX640933.1 | - | 3 | 575 | |||

ZNF600
Genomic Map