siRNA Details
Gene Symbol: ZNF497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207009.1 | NP_001193938.1 | 2 | 2627 | ||
NM_198458.2 | NP_940860.2 | 3 | 2724 | |||
GenBank mRNA | AK126727.1 | - | 3 | 2630 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207009.1 | NP_001193938.1 | 2 | 2627 | ||
NM_198458.2 | NP_940860.2 | 3 | 2724 | |||
GenBank mRNA | AK126727.1 | - | 3 | 2630 |