siRNA Details
| Gene Symbol: ZNF550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277090.1 | NP_001264019.1 | 4 | 1177 | ||
| NM_001277091.1 | NP_001264020.1 | 4 | 1177 | |||
| NM_001277092.1 | NP_001264021.1 | 4 | 1177 | |||
| XM_011526567.2 | XP_011524869.1 | 4 | 1174 | |||
| XM_011526568.2 | XP_011524870.1 | 4 | 2267 | |||
| XM_017026401.1 | XP_016881890.1 | 4 | 1174 | |||
| GenBank mRNA | AK122867.1 | - | 4 | 1178 | ||
| AK312802.1 | - | 5 | 1129 | |||
| AL833214.1 | - | 1 | 746 | |||
| BC053858.1 | - | 4 | 1151 | 6044705 | ||
| BX538032.1 | - | 4 | 1164 | |||

ZNF550
Genomic Map