siRNA Details
| Gene Symbol: OR7D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175883.2 | NP_787079.1 | 1 | 707 | ||
| GenBank mRNA | AK095468.1 | - | 1 | 707 | ||
| X64975.1 | - | 1 | 145 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR7D2| Gene Symbol: OR7D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175883.2 | NP_787079.1 | 1 | 707 | ||
| GenBank mRNA | AK095468.1 | - | 1 | 707 | ||
| X64975.1 | - | 1 | 145 | |||