siRNA Details
Gene Symbol: ZNF791 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153358.2 | NP_699189.2 | 4 | 465 | ||
XM_017026412.1 | XP_016881901.1 | 4 | 435 | |||
GenBank mRNA | AK074877.1 | - | 4 | 444 | ||
AK297715.1 | - | 3 | 302 | |||
AK298597.1 | - | 5 | 603 | |||
AK307775.1 | - | 3 | 404 | |||
BC106937.1 | - | 4 | 399 | 40033817 | ||
BC106938.1 | - | 4 | 398 | 40033819 |
