siRNA Details
| Gene Symbol: ZNF791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153358.2 | NP_699189.2 | 4 | 1261 | ||
| XM_017026412.1 | XP_016881901.1 | 4 | 1231 | |||
| GenBank mRNA | AK074877.1 | - | 4 | 1240 | ||
| AK297715.1 | - | 3 | 1098 | |||
| AK298597.1 | - | 5 | 1399 | |||
| AK307775.1 | - | 3 | 1200 | |||
| BC106937.1 | - | 4 | 1195 | 40033817 | ||
| BC106938.1 | - | 4 | 1194 | 40033819 | ||

ZNF791
Genomic Map