siRNA Details
| Gene Symbol: GBP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320257.1 | NP_001307186.1 | 9 | 1680 | ||
| NM_198460.2 | NP_940862.2 | 11 | 2021 | |||
| XM_011540835.2 | XP_011539137.1 | 11 | 1850 | |||
| GenBank mRNA | AK131329.1 | - | 6 | 903 | ||
| AK131356.1 | - | 11 | 2005 | |||
| AK299408.1 | - | 10 | 1656 | |||
| AK299466.1 | - | 9 | 1497 | |||
| AK303987.1 | - | 11 | 1765 | |||
| BC131713.1 | - | 11 | 1799 | 40122722 | ||
| BX537949.1 | - | 11 | 1840 | |||
| BX647907.1 | - | 11 | 1838 | |||

GBP6
Genomic Map