siRNA Details
| Gene Symbol: CITED4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133467.2 | NP_597724.1 | 1 | 995 | ||
| GenBank mRNA | BC035496.2 | - | 1 | 997 | 3622207 | |
| BC052559.1 | - | 1 | 995 | 6250908 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CITED4| Gene Symbol: CITED4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133467.2 | NP_597724.1 | 1 | 995 | ||
| GenBank mRNA | BC035496.2 | - | 1 | 997 | 3622207 | |
| BC052559.1 | - | 1 | 995 | 6250908 | ||