siRNA Details
| Gene Symbol: LEXM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110533.1 | NP_001104003.1 | 9 | 1172 | ||
| NM_152607.2 | NP_689820.2 | 9 | 1172 | |||
| XM_017000480.1 | XP_016855969.1 | 9 | 1172 | |||
| XM_017000481.1 | XP_016855970.1 | 9 | 1172 | |||
| GenBank mRNA | AK097520.1 | - | 9 | 1172 | ||
| BC039109.1 | - | 9 | 1133 | 4837126 | ||
| BC064142.1 | - | 9 | 1134 | 30390069 | ||

LEXM
Genomic Map