siRNA Details
| Gene Symbol: APCDD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304787.1 | NP_001291716.1 | 5 | 1681 | ||
| NM_153360.2 | NP_699191.1 | 4 | 1510 | |||
| GenBank mRNA | AK074647.1 | - | 4 | 1465 | ||
| AK293299.1 | - | 5 | 1640 | |||
| BC101758.1 | - | 4 | 1362 | 8069264 | ||
| BC101760.1 | - | 4 | 1362 | 8069266 | ||

APCDD1L
Genomic Map