siRNA Details
| Gene Symbol: CCDC116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152612.2 | NP_689825.2 | 5 | 1859 | ||
| XM_006724159.2 | XP_006724222.1 | 5 | 2078 | |||
| XM_011529984.2 | XP_011528286.1 | 4 | 2401 | |||
| XM_011529985.1 | XP_011528287.1 | 4 | 1496 | |||
| GenBank mRNA | AK093365.1 | - | 6 | 2205 | ||
| BC033499.1 | - | 5 | 1859 | 5172053 | ||

CCDC116
Genomic Map