siRNA Details
| Gene Symbol: LONRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198461.3 | NP_940863.3 | 8 | 2188 | ||
| GenBank mRNA | AK127206.1 | - | 8 | 890 | ||
| BC101662.1 | - | 7 | 847 | 8069168 | ||
| BC101664.1 | - | 7 | 847 | 8069170 | ||
| BC143468.1 | - | 7 | 847 | |||
| DQ020495.1 | - | 8 | 2188 | |||

LONRF2
Genomic Map