siRNA Details
| Gene Symbol: EMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001424.5 | NP_001415.1 | 5 | 565 | ||
| XM_006720864.3 | XP_006720927.1 | 5 | 559 | |||
| GenBank mRNA | AK313134.1 | - | 5 | 481 | ||
| AV657588.1 | - | 4 | 280 | |||
| BC009687.1 | - | 5 | 456 | 3895094 | ||
| BF981157.1 | - | 5 | 547 | |||
| U52100.1 | - | 4 | 400 | |||
| X94770.1 | - | 4 | 347 | |||

EMP2
Genomic Map