siRNA Details
| Gene Symbol: XIRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198621.2 | NP_001185550.1 | 2 | 2312 | ||
| NM_194293.2 | NP_919269.2 | 2 | 2302 | |||
| XM_005264909.3 | XP_005264966.1 | 2 | 2261 | |||
| GenBank mRNA | AJ626899.1 | - | 2 | 2310 | ||
| AJ626900.1 | - | 2 | 2310 | |||
| AK126299.1 | - | 1 | 752 | |||
| AL713648.1 | - | 1 | 390 | |||
| AL831956.1 | - | 1 | 97 | |||
| AL832020.1 | - | 1 | 390 | |||
| AL832459.1 | - | 1 | 390 | |||
| AY375160.1 | - | 2 | 2302 | |||
| CR749430.1 | - | 2 | 2312 | |||

XIRP1
Genomic Map