siRNA Details
| Gene Symbol: PRICKLE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198859.3 | NP_942559.1 | 8 | 2322 | ||
| XM_011533432.2 | XP_011531734.1 | 10 | 2206 | |||
| XM_011533433.2 | XP_011531735.1 | 9 | 2116 | |||
| XM_011533434.2 | XP_011531736.1 | 10 | 2031 | |||
| XM_011533435.2 | XP_011531737.1 | 9 | 1941 | |||
| XM_011533436.2 | XP_011531738.1 | 9 | 2264 | |||
| XM_011533437.2 | XP_011531739.1 | 9 | 11334 | |||
| XM_011533438.2 | XP_011531740.1 | 6 | 2156 | |||
| XM_011533440.2 | XP_011531742.1 | 10 | 2136 | |||
| XM_017005798.1 | XP_016861287.1 | 8 | 2046 | |||
| XM_017005799.1 | XP_016861288.1 | 7 | 3795 | |||
| GenBank mRNA | AK127113.1 | - | 1 | 337 | ||
| AL833539.1 | - | 8 | 2323 | |||
| BC040867.1 | - | 2 | 178 | |||
| BC119002.1 | - | 8 | 2303 | 40082599 | ||
| BX537915.1 | - | 2 | 150 | |||
| BX647797.1 | - | 3 | 1019 | |||
| CR936678.1 | - | 2 | 372 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001081146, NM_001134459, NM_001134461, NM_001134460, BC099897, and others.

PRICKLE2
Genomic Map