siRNA Details
| Gene Symbol: KBTBD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207335.2 | NP_997218.2 | 3 | 1445 | ||
| XM_005247167.1 | XP_005247224.1 | 4 | 1909 | |||
| XM_005247168.4 | XP_005247225.1 | 4 | 1987 | |||
| XM_011512490.2 | XP_011510792.1 | 4 | 1909 | |||
| XM_011512491.2 | XP_011510793.1 | 4 | 1909 | |||
| XM_011512492.2 | XP_011510794.1 | 4 | 1909 | |||
| GenBank mRNA | AK128176.1 | - | 3 | 307 | ||
| BX537865.1 | - | 3 | 484 | |||

KBTBD12
Genomic Map