siRNA Details
| Gene Symbol: DCLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040260.3 | NP_001035350.2 | 7 | 1908 | ||
| NM_001040261.4 | NP_001035351.4 | 8 | 1959 | |||
| NR_036614.1 | 7 | 1905 | ||||
| XM_005262785.2 | XP_005262842.1 | 8 | 1785 | |||
| XM_005262786.2 | XP_005262843.1 | 7 | 1734 | |||
| XM_017007832.1 | XP_016863321.1 | 8 | 1788 | |||
| XM_017007833.1 | XP_016863322.1 | 8 | 1788 | |||
| GenBank mRNA | AB209181.1 | - | 7 | 1908 | ||
| AL834498.1 | - | 8 | 1006 | |||
| BC032726.1 | - | 7 | 1198 | |||
| DA201927.1 | - | 5 | 355 | |||

DCLK2
Genomic Map