siRNA Details
| Gene Symbol: UGT3A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168316.1 | NP_001161788.1 | 3 | 768 | ||
| NM_174914.3 | NP_777574.2 | 4 | 870 | |||
| XM_011513988.2 | XP_011512290.1 | 5 | 912 | |||
| XM_017009150.1 | XP_016864639.1 | 4 | 921 | |||
| GenBank mRNA | AK075383.1 | - | 4 | 831 | ||
| AK300751.1 | - | 3 | 729 | |||
| AY358416.1 | - | 4 | 805 | |||
| BC103924.1 | - | 4 | 819 | 40007344 | ||
| BC103925.1 | - | 4 | 819 | 40007348 | ||

UGT3A2
Genomic Map