siRNA Details
| Gene Symbol: C6orf118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144980.3 | NP_659417.2 | 8 | 1350 | ||
| XM_005266838.2 | XP_005266895.1 | 8 | 1375 | |||
| XM_011535509.2 | XP_011533811.1 | 8 | 1537 | |||
| XM_011535510.2 | XP_011533812.1 | 7 | 1477 | |||
| XM_011535511.2 | XP_011533813.1 | 8 | 1537 | |||
| XM_011535512.2 | XP_011533814.1 | 8 | 1400 | |||
| XM_011535513.2 | XP_011533815.1 | 8 | 1803 | |||
| XM_017010324.1 | XP_016865813.1 | 8 | 2012 | |||
| GenBank mRNA | AK302358.1 | - | 8 | 1805 | ||
| BC026278.1 | - | 8 | 1337 | 4703212 | ||

C6orf118
Genomic Map