siRNA Details
| Gene Symbol: DDX53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182699.3 | NP_874358.2 | 1 | 1739 | ||
| GenBank mRNA | AY039237.1 | - | 1 | 1744 | ||
| BC051703.1 | - | 1724 | 5297066 | |||
| BC067878.1 | - | 1 | 1730 | 5295819 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DDX53| Gene Symbol: DDX53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182699.3 | NP_874358.2 | 1 | 1739 | ||
| GenBank mRNA | AY039237.1 | - | 1 | 1744 | ||
| BC051703.1 | - | 1724 | 5297066 | |||
| BC067878.1 | - | 1 | 1730 | 5295819 | ||