siRNA Details
| Gene Symbol: CNBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173538.2 | NP_775809.1 | 6 | 727 | ||
| XM_017013149.1 | XP_016868638.1 | 6 | 733 | |||
| GenBank mRNA | AK093121.1 | - | 6 | 730 | ||
| BC117326.1 | - | 6 | 727 | 40125877 | ||
| BC117328.1 | - | 6 | 727 | 40125879 | ||

CNBD1
Genomic Map