siRNA Details
| Gene Symbol: SLC30A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172811.1 | NP_001166282.1 | 7 | 848 | ||
| NM_001172813.1 | NP_001166284.1 | 8 | 1093 | |||
| NM_001172814.1 | NP_001166285.1 | 6 | 935 | |||
| NM_001172815.2 | NP_001166286.1 | 8 | 962 | |||
| NM_173851.2 | NP_776250.2 | 5 | 905 | |||
| GenBank mRNA | AK298294.1 | - | 9 | 1096 | ||
| AY117411.1 | - | 5 | 675 | |||
| BC126446.1 | - | 6 | 774 | |||
| BC143953.1 | - | 6 | 775 | |||
| EF560713.1 | - | 7 | 848 | |||

SLC30A8
Genomic Map