siRNA Details
| Gene Symbol: ZNF596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042415.2 | NP_001035880.1 | 6 | 949 | ||
| NM_001042416.2 | NP_001035881.1 | 6 | 1082 | |||
| NM_001287254.1 | NP_001274183.1 | 6 | 940 | |||
| NM_001287255.1 | NP_001274184.1 | 6 | 915 | |||
| NM_001287256.1 | NP_001274185.1 | 6 | 911 | |||
| NM_001287399.1 | NP_001274328.1 | 6 | 998 | |||
| NM_173539.3 | NP_775810.2 | 6 | 1000 | |||
| XM_011534744.1 | XP_011533046.1 | 6 | 922 | |||
| XM_017013166.1 | XP_016868655.1 | 5 | 2639 | |||
| GenBank mRNA | AK313454.1 | - | 6 | 823 | ||
| BC026190.1 | - | 6 | 823 | 4837032 | ||

ZNF596
Genomic Map