siRNA Details
| Gene Symbol: IDO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194294.2 | NP_919270.2 | 11 | 1440 | ||
| GenBank mRNA | AK092502.1 | - | 4 | 788 | ||
| AK128691.1 | - | 11 | 3069 | |||
| EF052681.1 | - | 10 | 1159 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IDO2| Gene Symbol: IDO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194294.2 | NP_919270.2 | 11 | 1440 | ||
| GenBank mRNA | AK092502.1 | - | 4 | 788 | ||
| AK128691.1 | - | 11 | 3069 | |||
| EF052681.1 | - | 10 | 1159 | |||