siRNA Details
| Gene Symbol: EN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001427.3 | NP_001418.2 | 2 | 967 | ||
| GenBank mRNA | BC104970.1 | - | 2 | 967 | 8143973 | |
| BC104972.1 | - | 2 | 967 | 8143975 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EN2| Gene Symbol: EN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001427.3 | NP_001418.2 | 2 | 967 | ||
| GenBank mRNA | BC104970.1 | - | 2 | 967 | 8143973 | |
| BC104972.1 | - | 2 | 967 | 8143975 | ||