siRNA Details
| Gene Symbol: FAM47B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152631.2 | NP_689844.2 | 1 | 1605 | ||
| GenBank mRNA | AK093101.1 | - | 1 | 1587 | ||
| BC035026.2 | - | 1 | 1605 | 4828379 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM47B| Gene Symbol: FAM47B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152631.2 | NP_689844.2 | 1 | 1605 | ||
| GenBank mRNA | AK093101.1 | - | 1 | 1587 | ||
| BC035026.2 | - | 1 | 1605 | 4828379 | ||