siRNA Details
| Gene Symbol: SUPT20HL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136234.1 | NP_001129706.1 | 1 | 497 | ||
| Gene Symbol: SUPT20HL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136233.1 | NP_001129705.1 | 1 | 497 | ||

SUPT20HL1
Genomic Map