siRNA Details
| Gene Symbol: CLEC4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130441.2 | NP_569708.1 | 4 | 715 | ||
| NM_203503.1 | NP_987099.1 | 3 | 622 | |||
| XM_017018940.1 | XP_016874429.1 | 3 | 354 | |||
| GenBank mRNA | AF293615.1 | - | 4 | 715 | ||
| AF325459.1 | - | 3 | 250 | |||
| AF325460.1 | - | 2 | 138 | |||
| AK122596.1 | - | 3 | 398 | |||
| AY358223.1 | - | 3 | 381 | |||
| BC074967.2 | - | 3 | 268 | 30915289 | ||
| BC074968.2 | - | 3 | 268 | 30915508 | ||
| BC102015.1 | - | 3 | 325 | 40030336 | ||
| BC102016.1 | - | 3 | 325 | 40030338 | ||
| BC102017.1 | - | 3 | 324 | 40030339 | ||
| BC114338.1 | - | 3 | 325 | 40030323 | ||

CLEC4C
Genomic Map