siRNA Details
Gene Symbol: PSORS1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014069.2 | NP_054788.2 | 2 | 554 | ||
GenBank mRNA | AB031480.1 | - | 2 | 545 | ||
AF484420.1 | - | 2 | 554 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PSORS1C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014069.2 | NP_054788.2 | 2 | 554 | ||
GenBank mRNA | AB031480.1 | - | 2 | 545 | ||
AF484420.1 | - | 2 | 554 |