siRNA Details
| Gene Symbol: ENO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001976.4 | NP_001967.3 | 5, 6 | 442 | ||
| NM_053013.3 | NP_443739.3 | 5, 6 | 400 | |||
| XM_005256521.2 | XP_005256578.1 | 5, 6 | 828 | |||
| XM_011523729.1 | XP_011522031.1 | 5, 6 | 418 | |||
| XM_017024346.1 | XP_016879835.1 | 5, 6 | 835 | |||
| GenBank mRNA | AK300662.1 | - | 5, 6 | 370 | ||
| AK309864.1 | - | 6, 7 | 493 | |||
| AU143784.1 | - | 5, 6 | 439 | |||
| BC017249.2 | - | 5, 6 | 357 | 4902149 | ||
| BJ995304.1 | - | 5, 6 | 416 | |||
| DA082822.1 | - | 5, 6 | 400 | |||
| DA566866.1 | - | 5, 6 | 398 | |||
| X16504.1 | - | 4, 5 | 304 | |||
| X51957.1 | - | 4, 5 | 316 | |||

ENO3
Genomic Map