siRNA Details
| Gene Symbol: ZNF431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319124.1 | NP_001306053.1 | 5 | 625 | ||
| NM_001319126.1 | NP_001306055.1 | 6 | 644 | |||
| NM_001319127.1 | NP_001306056.1 | 6 | 702 | |||
| NM_133473.2 | NP_597730.2 | 5 | 595 | |||
| XM_011527750.2 | XP_011526052.1 | 6 | 673 | |||
| GenBank mRNA | AB075849.1 | - | 5 | 556 | ||
| AK293072.1 | - | 5 | 592 | |||
| AK308127.1 | - | 5 | 600 | |||
| BC040506.1 | - | 5 | 547 | 5297447 | ||
| BX647597.1 | - | 6 | 630 | |||

ZNF431
Genomic Map