siRNA Details
| Gene Symbol: ENO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193503.1 | NP_001180432.1 | 6 | 804 | ||
| NM_001976.4 | NP_001967.3 | 8 | 975 | |||
| NM_053013.3 | NP_443739.3 | 8 | 933 | |||
| XM_005256521.2 | XP_005256578.1 | 8 | 1361 | |||
| XM_011523729.1 | XP_011522031.1 | 8 | 951 | |||
| XM_017024346.1 | XP_016879835.1 | 8 | 1368 | |||
| GenBank mRNA | AK300662.1 | - | 8 | 819 | ||
| AK300673.1 | - | 7 | 795 | |||
| AK300709.1 | - | 6 | 774 | |||
| AK309864.1 | - | 9 | 1026 | |||
| BC017249.2 | - | 8 | 890 | 4902149 | ||
| X16504.1 | - | 7 | 837 | |||
| X51957.1 | - | 7 | 849 | |||

ENO3
Genomic Map