siRNA Details
| Gene Symbol: ZNF675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138330.2 | NP_612203.2 | 4 | 467 | ||
| XM_017026442.1 | XP_016881931.1 | 5 | 580 | |||
| XM_017026443.1 | XP_016881932.1 | 5 | 1085 | |||
| XM_017026444.1 | XP_016881933.1 | 4 | 464 | |||
| GenBank mRNA | AB209601.1 | - | 1 | 2501 | ||
| AK093669.1 | - | 4 | 454 | |||
| AK307845.1 | - | 4 | 395 | |||
| AY044432.1 | - | 4 | 392 | |||
| DA777369.1 | - | 4 | 486 | |||

ZNF675
Genomic Map