siRNA Details
| Gene Symbol: FAM9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_205849.3 | NP_995321.1 | 6 | 661 | ||
| XM_011545464.2 | XP_011543766.1 | 13 | 1234 | |||
| GenBank mRNA | AF494344.1 | - | 6 | 660 | ||
| AK097501.1 | - | 5 | 605 | |||
| AK128470.1 | - | 6 | 3022 | |||
| BC120955.1 | - | 5 | 360 | 40115245 | ||
| BC120956.1 | - | 5 | 360 | 40115246 | ||

FAM9B
Genomic Map