siRNA Details
| Gene Symbol: FAM9C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174901.5 | NP_777561.1 | 2, 3 | 397 | ||
| XM_005274460.2 | XP_005274517.1 | 1, 2 | 138 | |||
| XM_017029321.1 | XP_016884810.1 | 1, 2 | 138 | |||
| GenBank mRNA | AF494345.1 | - | 2, 3 | 280 | ||
| AK313776.1 | - | 2, 3 | 234 | |||
| BC127957.1 | - | 1, 2 | 91 | 40014693 | ||
| BX647580.1 | - | 2, 3 | 202 | |||
| BX648111.1 | - | 2, 3 | 281 | |||
| BX648919.1 | - | 2, 3 | 282 | |||

FAM9C
Genomic Map