siRNA Details
| Gene Symbol: SPTSSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138288.3 | NP_612145.2 | 2 | 816 | ||
| GenBank mRNA | AK001993.1 | - | 2 | 797 | ||
| AL080066.1 | - | 2 | 797 | |||
| BC016805.1 | - | 2 | 798 | 4075637 | ||
| BC016808.1 | - | 2 | 798 | 4077762 | ||
| BC021701.1 | - | 2 | 797 | 4069530 | ||
| BC068480.1 | - | 2 | 910 | 5294549 | ||

SPTSSA
Genomic Map