siRNA Details
| Gene Symbol: B3GNT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138706.4 | NP_619651.3 | 2 | 1477 | ||
| GenBank mRNA | AK127544.1 | - | 2 | 1060 | ||
| AK172863.1 | - | 2 | 1402 | |||
| AK292773.1 | - | 2 | 1420 | |||
| BC025357.1 | - | 1 | 1018 | |||
| BC103908.1 | - | 2 | 1376 | 40005856 | ||
| BC103909.2 | - | 2 | 1376 | 40005863 | ||
| BC103910.1 | - | 2 | 1376 | 40005864 | ||

B3GNT6
Genomic Map