siRNA Details
| Gene Symbol: SDR16C5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318049.1 | NP_001304978.1 | 6 | 1480 | ||
| NM_001318050.1 | NP_001304979.1 | 5 | 1348 | |||
| NM_138969.3 | NP_620419.2 | 6 | 1480 | |||
| XM_011517479.1 | XP_011515781.1 | 5 | 1348 | |||
| GenBank mRNA | AB083038.1 | - | 6 | 1386 | ||
| AK057667.1 | - | 6 | 823 | |||
| AK095159.1 | - | 6 | 1481 | |||
| AK294634.1 | - | 5 | 751 | |||
| AY444559.1 | - | 5 | 747 | |||
| BC037219.1 | - | 6 | 827 | 5181472 | ||
| BC064525.1 | - | 6 | 799 | 5752917 | ||

SDR16C5
Genomic Map